Benzyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate 98%
Lieferant: Thermo Scientific
Synonyme:
Boc-Pyr-OBzl
H62912.06
H62912.06EA
369
EUR
H62912.06
Benzyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate 98%
Benzyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate
Purity: 98%
CAS No.: 113400-36-5
Molecular Formula: C17H21NO5
Formula Weight: 319.35
Physical Form: Powder
Appearance: White
MDL No.: MFCD04115781
Formel:
C₁₇H₂₁NO₅ Molecular Weight: 319,357 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115781 CAS-Nummer: 113400-36-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...