Boc-L-cyclopropylglycine 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-(tert-Butoxycarbonyl)-L-cyclopropylglycine, Boc-cyclopropyl-Gly-OH, (S)-2-[(tert-Butoxycarbonyl)amino]-2-cyclopropylacetic acid, N-Boc-L-cyclopropylglycine, (S)-(Boc-amino)-cyclopropylacetic acid, (S)-2-((tert-Butoxycarbonyl)amino)-2-cyclopropylacetic acid, N-Boc-2-cyclopropyl-L-glycine, (S)-2-(Boc-amino)-2-cyclopropylacetic acid
Purity: 95%
CAS No.: 155976-13-9
Molecular Formula: C10H17NO4
Formula Weight: 215.25
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Beilstein Reference: 6650020
Optical Rotation: +38°(c=1 in ethanol)
MDL No.: MFCD06659132
Formel:
C₁₀H₁₇NO₄ Lagertemperatur: Kühlschrank |
MDL:
MFCD06659132 CAS-Nummer: 155976-13-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...