trans-3-Aminocyclohexanecarbonitrile hydrochloride 97%

Lieferant: Thermo Fisher Scientific
Gefahr

H62938.03
H62938.03EA 438 EUR
H62938.03
trans-3-Aminocyclohexanecarbonitrile hydrochloride 97%
trans-3-Aminocyclohexanecarbonitrile hydrochloride

Purity: 97% CAS No.: 920966-30-9 Molecular Formula: C7H12N2.HCl Formula Weight: 160.64 UN No.: UN3439 MDL No.: MFCD22548413

Formel: C₇H₁₃ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22548413
CAS-Nummer: 920966-30-9
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR