Boc-D-4-Cyanophenylalanine 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Boc-D-Phe(4-CN)-OH, N-Boc-4-cyano-D-phenylalanine, Boc-4-cyano-D-Phe-OH
H62947.03
H62947.06
H62947.03EA
152
EUR
H62947.03
H62947.06
Boc-D-4-Cyanophenylalanine 98%
Boc-D-4-Cyanophenylalanine
Purity: 98%
CAS No.: 146727-62-0
Molecular Formula: C15H18N2O4
Formula Weight: 290.31
Beilstein Reference: 9339089
UN No.: UN3439
MDL No.: MFCD00672527
Formel:
C₁₅H₁₈N₂O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00672527 CAS-Nummer: 146727-62-0 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...