4-(Boc-aminomethyl)-3-fluorobenzeneboronic acid pinacol ester 96%

Lieferant: Thermo Fisher Scientific

Synonyme: 4-(Boc-aminomethyl)-3-fluorophenylboronic acid pinacol ester, 2-Methyl-2-propanyl [2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]carbamate, Multiple CAS found

H62948.03
H62948.03EA 312 EUR
H62948.03
4-(Boc-aminomethyl)-3-fluorobenzeneboronic acid pinacol ester 96%
4-(Boc-aminomethyl)-3-fluorobenzeneboronic acid pinacol ester

Purity: 96% Molecular Formula: C18H27BFNO4 Formula Weight: 351.22 Physical Form: Solid

Formel: C₁₈H₂₇BFNO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD22494714
CAS-Nummer: 1351501-44-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR