(R)-4-(1-aminoethyl)phenol 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H63001.03
H63001.06
H63001.03EA
191
EUR
H63001.03
H63001.06
(R)-4-(1-aminoethyl)phenol 97%
(R)-4-(1-aminoethyl)phenol
Purity: 97%
CAS No.: 134855-89-3
Molecular Formula: C8H11NO
Formula Weight: 137.18
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: White
UN No.: UN3259
MDL No.: MFCD03844647
Formel:
C₈H₁₁NO |
MDL:
MFCD03844647 CAS-Nummer: 134855-89-3 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...