3-Nitro-L-phenylalanine 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
(S)-2-Amino-3-(3-nitrophenyl)propanoic acid, H-Phe(3-NO₂)-OH
H63029.MD
H63029.03
H63029.06
H63029.MDEA
48.8
EUR
H63029.MD
H63029.03
H63029.06
3-Nitro-L-phenylalanine 95%
3-Nitro-L-phenylalanine
Purity: 95%
CAS No.: 19883-74-0
Molecular Formula: C9H10N2O4
Formula Weight: 210.19
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Off-white
MDL No.: MFCD01860661
Formel:
C₉H₁₀N₂O₄ |
MDL:
MFCD01860661 CAS-Nummer: 19883-74-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...