4,5-Difluor-2-nitrotoluol 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
1,2-Difluoro-4-methyl-5-nitrobenzene
H63030.06
H63030.06EA
75.6
EUR
H63030.06
4,5-Difluor-2-nitrotoluol 98%
4,5-Difluor-2-nitrotoluol
Purity: 98%
CAS No.: 127371-50-0
Molecular Formula: C7H5F2NO2
Formula Weight: 173.12
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colourless to pale yellow
UN No.: UN2810
MDL No.: MFCD04973772
Formel:
C₇H₅F₂NO₂ Molecular Weight: 173.12 g/mol |
MDL:
MFCD04973772 CAS-Nummer: 127371-50-0 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...