O-tert-Butyl-L-threonine 97%
Lieferant: Thermo Fisher Scientific
H63031.06
H63031.14
H63031.06EA
83.6
EUR
H63031.06
H63031.14
O-tert-Butyl-L-threonine 97%
O-tert-Butyl-L-threonine
Purity: 97%
CAS No.: 4378-13-6
Molecular Formula: C8H17NO3
Formula Weight: 175.23
Physical Form: Solid
Beilstein Reference: 2206127
Optical Rotation: -43°(c=1 in water)
MDL No.: MFCD00037766
Formel:
C₈H₁₇NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00037766 CAS-Nummer: 4378-13-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...