N-Fmoc-N-methyl-D-phenylalanine 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Fmoc-D-N-Me-Phe-OH, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-3-phenylpropanoic acid
H63055.03
H63055.06
H63055.03EA
158
EUR
H63055.03
H63055.06
N-Fmoc-N-methyl-D-phenylalanine 98%
N-Fmoc-N-methyl-D-phenylalanine
Purity: 98%
CAS No.: 138775-05-0
Molecular Formula: C25H23NO4
Formula Weight: 401.45
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Beilstein Reference: 8441627
MDL No.: MFCD00153392
Formel:
C₂₅H₂₃NO₄ Molecular Weight: 401.462 g/mol |
MDL:
MFCD00153392 CAS-Nummer: 138775-05-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...