DL-Glutaminsäure 95%

Lieferant: Thermo Fisher Scientific

H63075.14 H63075.22 H63075.36
H63075.14EA 69.6 EUR
H63075.14 H63075.22 H63075.36
DL-Glutaminsäure 95%
DL-Glutaminsäure

Purity: 95% CAS No.: 617-65-2 Molecular Formula: C5H9NO4 Formula Weight: 147.13 Storage Temperature: -30°C to -10°C Physical Form: Solid Beilstein Reference: 1723799 MDL No.: MFCD00063113

Formel: C₅H₉NO₄
Molecular Weight: 147.13 g/mol
MDL: MFCD00063113
CAS-Nummer: 617-65-2
EINECS: 210-522-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR