DL-Glutaminsäure 95%
Lieferant: Thermo Fisher Scientific
H63075.14
H63075.22
H63075.36
H63075.14EA
69.6
EUR
H63075.14
H63075.22
H63075.36
DL-Glutaminsäure 95%
DL-Glutaminsäure
Purity: 95%
CAS No.: 617-65-2
Molecular Formula: C5H9NO4
Formula Weight: 147.13
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Beilstein Reference: 1723799
MDL No.: MFCD00063113
Formel:
C₅H₉NO₄ Molecular Weight: 147.13 g/mol |
MDL:
MFCD00063113 CAS-Nummer: 617-65-2 EINECS: 210-522-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...