Z-Aib-OH 98%
Lieferant: Thermo Scientific
Synonyme:
N-Benzyloxycarbonyl-alpha-methylalanine, 2-(Benzyloxycarbonylamino)isobutyric acid, N-Benzyloxycarbonyl-2-methylalanine, Z-Aib-OH, 2-(Cbz-amino)isobutyric acid, 2-(Carbobenzoxyamino)isobutyric acid, N-Cbz-alpha-methylalanine
H63119.06
H63119.06EA
74.5
EUR
H63119.06
Z-Aib-OH 98%
Z-Aib-OH
Purity: 98%
CAS No.: 15030-72-5
Molecular Formula: C12H15NO4
Formula Weight: 237.25
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Melting Point: 65-69° dec
Beilstein Reference: 2218009
MDL No.: MFCD00004190
Formel:
C₁₂H₁₅NO₄ Molecular Weight: 237,255 g/mol Schmelzpunkt: 65…69 °C (Zers.) |
MDL:
MFCD00004190 CAS-Nummer: 15030-72-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...