DL-m-Tyrosin 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Amino-3-(3-hydroxyphenyl)propionsäure, 3-Hydroxyphenylalanin, 3-(m-Hydroxyphenyl)-DL-alanin
H63163.06
H63163.06EA
216
EUR
H63163.06
DL-m-Tyrosin 98%
DL-m-Tyrosin
Purity: 98%
CAS No.: 775-06-4
Molecular Formula: C9H11NO3
Formula Weight: 181.19
Physical Form: Powder
Appearance: Off-white to yellow
Melting Point: 280-285° dec
Beilstein Reference: 2416853
MDL No.: MFCD00002597
Formel:
C₉H₁₁NO₃ Schmelzpunkt: 280…285 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002597 CAS-Nummer: 775-06-4 EINECS: 212-270-9 Merck Index: 14,9840 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...