Fmoc-N-Me-Thr(tBu)-OH 99%
Lieferant: Thermo Scientific
Synonyme:
(2S,3R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-3-(tert-butoxy)butanoic acid, N-Fmoc-O-tert-butyl-N-methyl-L-threonine
H63170.03
H63170.03EA
289
EUR
H63170.03
Fmoc-N-Me-Thr(tBu)-OH 99%
Fmoc-N-Me-Thr(tBu)-OH
Purity: 99%
CAS No.: 117106-20-4
Molecular Formula: C24H29NO5
Formula Weight: 411.49
Storage Temperature: 2°C to 8°C
Physical Form: Solid
MDL No.: MFCD02094431
Formel:
C₂₄H₂₉NO₅ Molecular Weight: 411,498 g/mol |
MDL:
MFCD02094431 CAS-Nummer: 117106-20-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...