Boc-D-Glu(OtBu)-OH 98%

Lieferant: Thermo Scientific

Synonyme: N-Boc-D-glutamic acid 5-tert-butyl ester

H63172.06
H63172.06EA 81.4 EUR
H63172.06
Boc-D-Glu(OtBu)-OH 98%
Boc-D-Glu(OtBu)-OH

Purity: 98% CAS No.: 104719-63-3 Molecular Formula: C14H25NO6 Formula Weight: 303.35 Physical Form: Solid Beilstein Reference: 3594191 MDL No.: MFCD00076927

Formel: C₁₄H₂₅NO₆
Molecular Weight: 303,355 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00076927
CAS-Nummer: 104719-63-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR