L-Glutaminsäure-5-tert.-butylester 98%

Lieferant: Thermo Scientific

Synonyme: H-Glu(OtBu)-OH, L-Glutamic Acid 5-tert-Butyl Ester

H63223.06
H63223.06EA 148 EUR
H63223.06
L-Glutaminsäure-5-tert.-butylester 98%
5-tert-Butyl-L-glutamate hydrate

Purity: 98% CAS No.: 2419-56-9 Molecular Formula: C9H17NO4 Formula Weight: 203.24 Storage Temperature: 2°C to 8°C Physical Form: Solid Beilstein Reference: 2049031 MDL No.: MFCD00038580

Formel: C₉H₁₇NO₄
Molecular Weight: 203,24 g/mol
MDL: MFCD00038580
CAS-Nummer: 2419-56-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR