4-(Brommethyl)-3-fluorbenzonitril 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 95%
CAS No.: 105942-09-4
Molecular Formula: C8H5BrFN
Formula Weight: 214.04
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: Off-white
Melting Point: 75-79° dec
Beilstein Reference: 7758134
UN No.: UN2923
MDL No.: MFCD07368341
Formel:
C₈H₅BrFN |
MDL:
MFCD07368341 CAS-Nummer: 105942-09-4 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...