Boc-N-methyl-L-serine 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Boc-N-Me-Ser-OH, N-Boc-N-methyl-L-serine
H63314.03
H63314.03EA
101
EUR
H63314.03
Boc-N-methyl-L-serine 98%
Boc-N-methyl-L-serine
Purity: 98%
CAS No.: 101772-29-6
Molecular Formula: C9H17NO5
Formula Weight: 219.23
Physical Form: Solid
Melting Point: 83-87°
Beilstein Reference: 8685334
Optical Rotation: -33°(c=1 in methanol)
MDL No.: MFCD00037249
Formel:
C₉H₁₇NO₅ Schmelzpunkt: 83…87 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00037249 CAS-Nummer: 101772-29-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...