2-(Trifluormethyl)isonicotinsäure 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-(Trifluoromethyl)pyridine-4-carboxylic acid

H63315.03
H63315.03EA 212 EUR
H63315.03
2-(Trifluormethyl)isonicotinsäure 97%
2-(Trifluoromethyl)isonicotinic acid

Purity: 97% CAS No.: 131747-41-6 Molecular Formula: C7H4F3NO2 Formula Weight: 191.11 Physical Form: Solid Appearance: Off-white Melting Point: 217-223° MDL No.: MFCD07774132

Formel: C₇H₄F₃NO₂
Molecular Weight: 191,11 g/mol
Schmelzpunkt: 217…223 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07774132
CAS-Nummer: 131747-41-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR