(R)-tert-Butyl-2-amino-3-phenylpropanoate hydrochloride 98%

Lieferant: Thermo Fisher Scientific

Synonyme: D-Phenylalanine tert-butyl ester hydrochloride, H-D-Phe-OtBu·HCl, H-D-Phe-OtBu.HCl

H63325.03 H63325.06
H63325.03EA 30.1 EUR
H63325.03 H63325.06
(R)-tert-Butyl-2-amino-3-phenylpropanoate hydrochloride 98%
(R)-tert-Butyl-2-amino-3-phenylpropanoate hydrochloride

Purity: 98% CAS No.: 3403-25-6 Molecular Formula: C13H19NO2.HCl Formula Weight: 257.76 Storage Temperature: -30°C to -10°C Physical Form: Solid MDL No.: MFCD00153455

Formel: C₁₃H₁₉NO₂·HCl
Molecular Weight: 257.8 g/mol
MDL: MFCD00153455
CAS-Nummer: 3403-25-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR