(2-Chlor-3-pyridinyl)methanol 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 97%
CAS No.: 42330-59-6
Molecular Formula: C6H6ClNO
Formula Weight: 143.57
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 61-65° dec
Boiling Point: 112°/0.2mm
Beilstein Reference: 1525378
MDL No.: MFCD01935961
Formel:
C₆H₆ClNO Siedepunkt: 112 °C (0,2 mmHg) Schmelzpunkt: 61…65 °C (Zersetzung) |
MDL:
MFCD01935961 CAS-Nummer: 42330-59-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...