Z-D-Glu(OtBu)-OH 98%
Lieferant: Thermo Fisher Scientific
H63371.06
H63371.06EA
87.8
EUR
H63371.06
Z-D-Glu(OtBu)-OH 98%
Z-D-Glu(OtBu)-OH
Purity: 98%
CAS No.: 51644-83-8
Molecular Formula: C17H23NO6
Formula Weight: 337.37
Physical Form: Solid
MDL No.: MFCD00077018
Formel:
C₁₇H₂₃NO₆ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00077018 CAS-Nummer: 51644-83-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...