Methyl-2-methylpropionimidate hydrochloride 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H63386.03
H63386.06
H63386.03EA
59
EUR
H63386.03
H63386.06
Methyl-2-methylpropionimidate hydrochloride 97%
Methyl-2-methylpropionimidate hydrochloride
Purity: 97%
CAS No.: 39739-60-1
Molecular Formula: C5H11NO.HCl
Formula Weight: 137.61
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: White
MDL No.: MFCD03844704
Formel:
C₅H₁₂ClNO |
MDL:
MFCD03844704 CAS-Nummer: 39739-60-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...