Indol-4-carbonitril ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 4-Indolcarbonitril, 4-Cyanindol, 1H-Indol-4-carbonitril, 4-Cyan-1H-indol

H63391.03 H63391.06
H63391.03EA 34.5 EUR
H63391.03 H63391.06
Indol-4-carbonitril ≥99%
Indol-4-carbonitril

Purity: 99% CAS No.: 16136-52-0 Molecular Formula: C9H6N2 Formula Weight: 142.16 Storage Temperature: -30°C to -10°C Physical Form: Solid Melting Point: 120-125° MDL No.: MFCD00152045

Formel: C₉H₆N₂
Molecular Weight: 142.16 g/mol
Schmelzpunkt: 120…125 °C
MDL: MFCD00152045
CAS-Nummer: 16136-52-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR