(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)propanoic acid 95%

Lieferant: Thermo Fisher Scientific

Synonyme: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-alanine, Fmoc-N-Me-Ala-OH, N-Fmoc-N-methyl-L-alanine

H63411.06 H63411.14
H63411.06EA 86.5 EUR
H63411.06 H63411.14
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)propanoic acid 95%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)propanoic acid

Purity: 95% CAS No.: 84000-07-7 Molecular Formula: C19H19NO4 Formula Weight: 325.36 Storage Temperature: 2°C to 8°C Physical Form: Solid Melting Point: 140° Beilstein Reference: 4329998 Optical Rotation: -18°(c=1 in DMF) MDL No.: MFCD00153384

Formel: C₁₉H₁₉NO₄
Molecular Weight: 325.4 g/mol
MDL: MFCD00153384
CAS-Nummer: 84000-07-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR