4-(Trifluoromethylthio)phenylacetic acid 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-(4-((Trifluoromethyl)thio)phenyl)acetic acid, 4-(Carboxymethyl)phenyl trifluoromethyl sulphide, {4-[(Trifluoromethyl)sulphanyl]phenyl}acetic acid, 4-[(Trifluoromethyl)sulphanyl]phenylacetic acid

H63432.03 H63432.06
H63432.03EA 60.8 EUR
H63432.03 H63432.06
4-(Trifluoromethylthio)phenylacetic acid 98%
4-(Trifluoromethylthio)phenylacetic acid

Purity: 98% CAS No.: 243977-23-3 Molecular Formula: C9H7F3O2S Formula Weight: 236.21 Physical Form: Solid MDL No.: MFCD00236351

Formel: C₉H₇F₃O₂S
Molecular Weight: 236.208 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00236351
CAS-Nummer: 243977-23-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR