4-(Trifluoromethylthio)phenylacetic acid 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-(4-((Trifluoromethyl)thio)phenyl)acetic acid, 4-(Carboxymethyl)phenyl trifluoromethyl sulphide, {4-[(Trifluoromethyl)sulphanyl]phenyl}acetic acid, 4-[(Trifluoromethyl)sulphanyl]phenylacetic acid
H63432.03
H63432.06
H63432.03EA
60.8
EUR
H63432.03
H63432.06
4-(Trifluoromethylthio)phenylacetic acid 98%
4-(Trifluoromethylthio)phenylacetic acid
Purity: 98%
CAS No.: 243977-23-3
Molecular Formula: C9H7F3O2S
Formula Weight: 236.21
Physical Form: Solid
MDL No.: MFCD00236351
Formel:
C₉H₇F₃O₂S Molecular Weight: 236.208 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236351 CAS-Nummer: 243977-23-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...