2,6-Dichlornicotinonitril ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H63480.03
H63480.06
H63480.03EA
84.9
EUR
H63480.03
H63480.06
2,6-Dichlornicotinonitril ≥97%
2,6-Dichlornicotinonitril
Formel:
C₆H₂Cl₂N₂ Schmelzpunkt: 121…125 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03411341 CAS-Nummer: 40381-90-6 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...