(R)-3-([1,1'-Biphenyl]-4-yl)-2-((tert-butoxycarbonyl)amino)propanoic acid 95%

Lieferant: Thermo Fisher Scientific

Synonyme: Boc-4-phenyl-D-Phe-OH, N-Boc-3-(4-biphenylyl)-D-alanine

H63494.06
H63494.06EA 201 EUR
H63494.06
(R)-3-([1,1'-Biphenyl]-4-yl)-2-((tert-butoxycarbonyl)amino)propanoic acid 95%
(R)-3-([1,1'-Biphenyl]-4-yl)-2-((tert-butoxycarbonyl)amino)propanoic acid
Formel: C₂₀H₂₃NO₄
MDL: MFCD00191185
CAS-Nummer: 128779-47-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR