Ethyl-3-(2-bromophenyl)propanoate 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Ethyl 3-(2-bromophenyl)propanoate

H63505.06
H63505.06EA 108 EUR
H63505.06
Ethyl-3-(2-bromophenyl)propanoate 98%
Ethyl-3-(2-bromophenyl)propanoate

Purity: 98% CAS No.: 135613-33-1 Molecular Formula: C11H13BrO2 Formula Weight: 257.13 Physical Form: Liquid Appearance: Pale yellow MDL No.: MFCD07772962

Formel: C₁₁H₁₃BrO₂
Molecular Weight: 257,127 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07772962
CAS-Nummer: 135613-33-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR