3-(4-Thiazolyl)-L-alanine 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: H-3-(4-Thiazolyl)-Ala-OH, (S)-2-Amino-3-(thiazol-4-yl)propanoic acid

H63511.03
H63511.03EA 380 EUR
H63511.03
3-(4-Thiazolyl)-L-alanine 95%
3-(4-Thiazolyl)-L-alanine

Purity: 95% CAS No.: 119433-80-6 Molecular Formula: C6H8N2O2S Formula Weight: 172.20 Storage Temperature: -30°C to -10°C Physical Form: Solid Appearance: Pale yellow MDL No.: MFCD00079677

Formel: C₆H₈N₂O₂S
Molecular Weight: 172.202 g/mol
MDL: MFCD00079677
CAS-Nummer: 119433-80-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR