2,4-Dimethoxy-1-fluorobenzene 97%

Lieferant: Thermo Fisher Scientific
Achtung

H63528.03 H63528.06
H63528.03EA 94.3 EUR
H63528.03 H63528.06
2,4-Dimethoxy-1-fluorobenzene 97%
2,4-Dimethoxy-1-fluorobenzene

Purity: 97% CAS No.: 17715-70-7 Molecular Formula: C8H9FO2 Formula Weight: 156.15 Flash Point: 105°(221°F) Physical Form: Liquid Density: 1.188 Appearance: Pale yellow MDL No.: MFCD00042265

Formel: C₈H₉FO₂
Dichte: 1,188
Flammpunkt: 105 °C (221 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00042265
CAS-Nummer: 17715-70-7
EINECS: 605-794-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR