Methyl-2-brom-1,3-thiazol-4-carboxylat 96%

Lieferant: Thermo Scientific

Synonyme: Methyl 2-bromothiazole-4-carboxylate

H63541.06
H63541.06EA 201 EUR
H63541.06
Methyl-2-brom-1,3-thiazol-4-carboxylat 96%
Methyl-2-bromothiazole-4-carboxylate

Purity: 96% CAS No.: 170235-26-4 Molecular Formula: C5H4BrNO2S Formula Weight: 222.06 Physical Form: Solid MDL No.: MFCD06659908

Formel: C₅H₄BrNO₂S
Molecular Weight: 222,06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659908
CAS-Nummer: 170235-26-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR