4-tert-Butyl-N-(tert-butoxycarbonyl)-L-aspartate 98%

Lieferant: Thermo Fisher Scientific

Synonyme: 4-tert-Butyl N-Boc-L-aspartate, N-(tert-Butoxycarbonyl)-L-aspartic Acid 4-tert-Butyl Ester, N-Boc-L-aspartic Acid 4-tert-Butyl Ester, Boc-Asp(OtBu)-OH

H63554.06 H63554.14
H63554.06EA 61.5 EUR
H63554.06 H63554.14
4-tert-Butyl-N-(tert-butoxycarbonyl)-L-aspartate 98%
4-tert-Butyl-N-(tert-butoxycarbonyl)-L-aspartate

Purity: 98% CAS No.: 1676-90-0 Molecular Formula: C13H23NO6 Formula Weight: 289.32 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Melting Point: 64-67° Beilstein Reference: 2336144 Optical Rotation: +2°(c=1 in methanol) MDL No.: MFCD00076912

Formel: C₁₃H₂₃NO₆
Schmelzpunkt: 64…67 °C
MDL: MFCD00076912
CAS-Nummer: 1676-90-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR