4-tert-Butyl-N-(tert-butoxycarbonyl)-L-aspartate 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
4-tert-Butyl N-Boc-L-aspartate, N-(tert-Butoxycarbonyl)-L-aspartic Acid 4-tert-Butyl Ester, N-Boc-L-aspartic Acid 4-tert-Butyl Ester, Boc-Asp(OtBu)-OH
H63554.06
H63554.14
H63554.06EA
61.5
EUR
H63554.06
H63554.14
4-tert-Butyl-N-(tert-butoxycarbonyl)-L-aspartate 98%
4-tert-Butyl-N-(tert-butoxycarbonyl)-L-aspartate
Purity: 98%
CAS No.: 1676-90-0
Molecular Formula: C13H23NO6
Formula Weight: 289.32
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Melting Point: 64-67°
Beilstein Reference: 2336144
Optical Rotation: +2°(c=1 in methanol)
MDL No.: MFCD00076912
Formel:
C₁₃H₂₃NO₆ Schmelzpunkt: 64…67 °C |
MDL:
MFCD00076912 CAS-Nummer: 1676-90-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...