2-Chloro-1-methylbenzimidazole

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Chloro-1-methyl-1H-benzo[d]imidazole

H63559.03
H63559.03EA 107 EUR
H63559.03
2-Chloro-1-methylbenzimidazole
2-Chloro-1-methylbenzimidazole

CAS No.: 1849-02-1 Molecular Formula: C8H7ClN2 Formula Weight: 166.61 Physical Form: Solid Appearance: White to yellow MDL No.: MFCD00769821

Formel: C₈H₇ClN₂
Molecular Weight: 166,608 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00769821
CAS-Nummer: 1849-02-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR