Ethyl-6-nitro-1H-indole-2-carboxylate 97%
Lieferant: Thermo Scientific
Synonyme:
Ethyl 6-nitroindole-2-carboxylate, Ethyl 6-nitro-1H-indole-2-carboxylate
H63569.03
H63569.03EA
290
EUR
H63569.03
Ethyl-6-nitro-1H-indole-2-carboxylate 97%
Ethyl-6-nitro-1H-indole-2-carboxylate
Purity: 97%
CAS No.: 16792-45-3
Molecular Formula: C11H10N2O4
Formula Weight: 234.21
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: Yellow
MDL No.: MFCD04973967
Formel:
C₁₁H₁₀N₂O₄ Molecular Weight: 234,211 g/mol |
MDL:
MFCD04973967 CAS-Nummer: 16792-45-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...