Fmoc-Asp(OAll)-OH 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Fmoc-Asp(allyl ester)-OH, N-Fmoc-L-aspartic acid 4-allyl ester
H63585.06
H63585.14
H63585.06EA
100
EUR
H63585.06
H63585.14
Fmoc-Asp(OAll)-OH 98%
Fmoc-Asp(OAll)-OH
Purity: 98%
CAS No.: 146982-24-3
Molecular Formula: C22H21NO6
Formula Weight: 395.41
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 111-115°
Beilstein Reference: 6670219
Optical Rotation: -27°(c=1 in DMF)
MDL No.: MFCD00190874
Formel:
C₂₂H₂₁NO₆ Schmelzpunkt: 111…115 °C |
MDL:
MFCD00190874 CAS-Nummer: 146982-24-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...