N-Boc-thiourea 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H63599.03
H63599.06
H63599.03EA
88.6
EUR
H63599.03
H63599.06
N-Boc-thiourea 97%
N-Boc-thiourea
Purity: 97%
CAS No.: 268551-65-1
Molecular Formula: C6H12N2O2S
Formula Weight: 176
Physical Form: Powder
Appearance: Off-white
Melting Point: 142° dec
MDL No.: MFCD04039987
Formel:
C₆H₁₂N₂O₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039987 CAS-Nummer: 268551-65-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...