5-Amino-3-methyl-1-phenyl-1H-pyrazol 99+%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Methyl-1-phenyl-1H-pyrazol-5-amine, 5-Amino-3-methyl-1-phenylpyrazole, 3-methyl-1-phenyl-1H-pyrazol-5-amine

H63605.06
H63605.06EA 51.6 EUR
H63605.06
5-Amino-3-methyl-1-phenyl-1H-pyrazol 99+%
5-Amino-3-methyl-1-phenyl-1H-pyrazole

Purity: 99+% CAS No.: 1131-18-6 Molecular Formula: C10H11N3 Formula Weight: 173.21 Physical Form: Solid Appearance: Yellow to pale brown Melting Point: 114-117° Beilstein Reference: 140666 MDL No.: MFCD00020727

Formel: C₁₀H₁₁N₃
Molecular Weight: 173.21 g/mol
Schmelzpunkt: 114…117 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00020727
CAS-Nummer: 1131-18-6
EINECS: 214-463-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR