Fmoc-N-Me-Ser(tBu)-OH 97%
Lieferant: Thermo Scientific
Synonyme:
N-Fmoc-O-tert-butyl-N-methyl-L-serine
H63614.06
H63614.06EA
521
EUR
H63614.06
Fmoc-N-Me-Ser(tBu)-OH 97%
Fmoc-N-Me-Ser(tBu)-OH
Purity: 97%
CAS No.: 197632-77-2
Molecular Formula: C23H27NO5
Formula Weight: 397.46
Physical Form: Crystalline solid
Appearance: White to off-white
Melting Point: 213-218°
MDL No.: MFCD02094430
Formel:
C₂₃H₂₇NO₅ Schmelzpunkt: 213…218 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02094430 CAS-Nummer: 197632-77-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...