2-Bromo-3-fluorobenzyl bromide 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-Bromo-1-(bromomethyl)-3-fluorobenzene

H63632.06 H63632.03
H63632.06EA 181 EUR
H63632.06 H63632.03
2-Bromo-3-fluorobenzyl bromide 98%
2-Bromo-3-fluorobenzyl bromide

Purity: 98% CAS No.: 1184918-22-6 Molecular Formula: C7H5Br2F Formula Weight: 267.92 UN No.: UN3261 MDL No.: MFCD12911576

Formel: C₇H₅Br₂F
Lagertemperatur: Raumtemperatur
MDL: MFCD12911576
CAS-Nummer: 1184918-22-6
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR