2-Bromo-3-fluorobenzyl bromide 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-Bromo-1-(bromomethyl)-3-fluorobenzene
H63632.06
H63632.03
H63632.06EA
181
EUR
H63632.06
H63632.03
2-Bromo-3-fluorobenzyl bromide 98%
2-Bromo-3-fluorobenzyl bromide
Purity: 98%
CAS No.: 1184918-22-6
Molecular Formula: C7H5Br2F
Formula Weight: 267.92
UN No.: UN3261
MDL No.: MFCD12911576
Formel:
C₇H₅Br₂F Lagertemperatur: Raumtemperatur |
MDL:
MFCD12911576 CAS-Nummer: 1184918-22-6 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...