Fmoc-N-Me-Ile-OH 95%

Lieferant: Thermo Scientific

Synonyme: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-isoleucine, Fmoc-N-Me-Ile-OH, N-Fmoc-N-methyl-L-isoleucine

H63640.06
H63640.06EA 301 EUR
H63640.06
Fmoc-N-Me-Ile-OH 95%
Fmoc-N-Me-Ile-OH

Purity: 95% CAS No.: 138775-22-1 Molecular Formula: C22H25NO4 Formula Weight: 367.44 Storage Temperature: 2°C to 8°C Physical Form: Solid Beilstein Reference: 7548444 Optical Rotation: -59°(c=1 in DMF) MDL No.: MFCD00153389

Formel: C₂₂H₂₅NO₄
Molecular Weight: 367,445 g/mol
MDL: MFCD00153389
CAS-Nummer: 138775-22-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR