Z-Asp(OMe)-OH 98%

Lieferant: Thermo Fisher Scientific

H63642.06
H63642.06EA 78 EUR
H63642.06
Z-Asp(OMe)-OH 98%
Z-Asp(OMe)-OH

Purity: 98% CAS No.: 3160-47-2 Molecular Formula: C13H15NO6 Formula Weight: 281.26 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Beilstein Reference: 6279422 MDL No.: MFCD00133585

Formel: C₁₃H₁₅NO₆
MDL: MFCD00133585
CAS-Nummer: 3160-47-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR