7-Chloro-1,2,3,4-tetrahydroisoquinoline 95%

Lieferant: Thermo Scientific
Achtung

H63693.03
H63693.03EA 677 EUR
H63693.03
7-Chloro-1,2,3,4-tetrahydroisoquinoline 95%
7-Chloro-1,2,3,4-tetrahydroisoquinoline

Purity: 95% CAS No.: 82771-60-6 Molecular Formula: C9H10ClN Formula Weight: 167.64 Storage Temperature: -30°C to -10°C Physical Form: Solid MDL No.: MFCD05861543

Formel: C₉H₁₀ClN
Molecular Weight: 167,636 g/mol
MDL: MFCD05861543
CAS-Nummer: 82771-60-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR