Di-tert-butyl L-aspartate hydrochloride 98%

Lieferant: Thermo Fisher Scientific

H63716.14
H63716.14EA 312 EUR
H63716.14
Di-tert-butyl L-aspartate hydrochloride 98%
Di-tert-butyl L-aspartate hydrochloride

Purity: 98% CAS No.: 1791-13-5 Molecular Formula: C12H23NO4.HCl Formula Weight: 281.78 Storage Temperature: -30°C to -10°C Physical Form: Solid Beilstein Reference: 5652807 MDL No.: MFCD00034851

Formel: C₁₂H₂₃NO₄
MDL: MFCD00034851
CAS-Nummer: 1791-13-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR