(R)-1-[α-(4-Chlorophenyl)benzyl]piperazine 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Chlorobenzhydryl Piperazine, (R)-1-((4-Chlorophenyl)(phenyl)methyl)piperazine
H63722.06
H63722.06EA
400
EUR
H63722.06
(R)-1-[α-(4-Chlorophenyl)benzyl]piperazine 97%
(R)-1-[alpha-(4-Chlorophenyl)benzyl]piperazine
Purity: 97%
CAS No.: 300543-56-0
Molecular Formula: C17H19ClN2
Formula Weight: 286.8
Physical Form: Solid
Appearance: White
MDL No.: MFCD11519277
Formel:
C₁₇H₁₉ClN₂ Molecular Weight: 286,803 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11519277 CAS-Nummer: 300543-56-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...