H-Lys(boc)-OMe·HCl 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
H-Lys(Boc)-OMe·HCl, Nepsilon-Boc-L-lysine methyl ester hydrochloride, H-Lys(Boc)-OMe.HCl
H63727.03
H63727.06
H63727.03EA
59.2
EUR
H63727.03
H63727.06
H-Lys(boc)-OMe·HCl 98%
H-Lys(boc)-OMe·HCl
Purity: 98%
CAS No.: 2389-48-2
Molecular Formula: C12H24N2O4.HCl
Formula Weight: 296.79
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Beilstein Reference: 5173577
MDL No.: MFCD00076959
Formel:
C₁₂H₂₅ClN₂O₄ |
MDL:
MFCD00076959 CAS-Nummer: 2389-48-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...